Learn More
Methyldiphenylsilane, 97%
Product Code. 11376846
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376846 25 g 25g
11366846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11376846 Supplier Thermo Scientific Alfa Aesar Supplier No. L04211.14
 Restricted Item
På lager
Add to basket
CAS: 776-76-1 | C13H13Si | 197.332 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 776-76-1
Molecular Formula C13H13Si
Molecular Weight (g/mol) 197.332
MDL Number MFCD00008255
InChI Key OKHRRIGNGQFVEE-UHFFFAOYSA-N
Synonym methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-,
PubChem CID 6327660
IUPAC Name methyl(diphenyl)silicon
SMILES C[Si](C1=CC=CC=C1)C2=CC=CC=C2
Specifications
Boiling Point 266°C to 267°C
Quantity 25 g
Flash Point 120°C (248°F)
Odor Odorless
Refractive Index 1.572
Density 1.001
Beilstein 1867183
Formula Weight 198.34
Percent Purity 97%
Chemical Name or Material Methyldiphenylsilane