missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10234010
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
This item is not returnable. View return policy
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
Melting Point -126°C
Color Colorless
Boiling Point 101°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 98.19g/mol
Physical Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylcyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'