Få mere at vide
Methylboronic Acid-d3, TRC
Artikelnummer. 30468619
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30468619 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468619 Leverandør Toronto Research Chemicals Leverandørnr. TRCM265092100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methylboronic Acid-d3
CAS 1332481-37-4
Molekylær formel CH2D3BO2
Mængde 100 mg
Synonym Methaneboronic Acid-d3, B-(Methyl-d3)boronoic Acid
InChI formel InChI=1S/CH5BO2/c1-2(3)4/h3-4H,1H3/i1D3
SMIL [2H]C([2H])([2H])B(O)O
IUPAC navn (methyl-d3)boronic acid
Molekylvægt (g/mol) 62.88
Formel vægt 63.06