Få mere at vide
Methylated Hesperidin (mixture of isomers), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylated Hesperidin (mixture of isomers) |
| CAS | 11013-97-1 |
| Molekylær formel | C29H36O15 |
| Mængde | 250 mg |
| Synonym | (S)-7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,3-dihydro-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one Monomethyl Ether, Methyl HES, (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one Compound With Methanol (1:1) |
| InChI formel | InChI=1S/C29H36O15/c1-11-22(32)24(34)26(36)28(41-11)40-10-20-23(33)25(35)27(37)29(44-20)42-13-7-14(30)21-15(31)9-17(43-19(21)8-13)12-4-5-16(38-2)18(6-12)39-3/h4-8,11,17,20,22-30,32-37H,9-10H2,1-3H3/t11-,17-,20+,22-,23+,24+,25-,26+,27+,28+,29+/m0/s1 |
| SMIL | COc1ccc(cc1OC)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2 |
| Molekylvægt (g/mol) | 624.59 |