missing translation for 'onlineSavingsMsg'
Få mere at vide

Methylated Hesperidin (mixture of isomers), TRC

Product Code. 30469824
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30469824 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469824 Supplier Toronto Research Chemicals Supplier No. TRCM31209725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methylated Hesperidin (mixture of isomers)
CAS 11013-97-1
Molekylær formel C29H36O15
Mængde 25 mg
Synonym (S)-7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,3-dihydro-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one Monomethyl Ether, Methyl HES, (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one Compound With Methanol (1:1)
InChI formel InChI=1S/C29H36O15/c1-11-22(32)24(34)26(36)28(41-11)40-10-20-23(33)25(35)27(37)29(44-20)42-13-7-14(30)21-15(31)9-17(43-19(21)8-13)12-4-5-16(38-2)18(6-12)39-3/h4-8,11,17,20,22-30,32-37H,9-10H2,1-3H3/t11-,17-,20+,22-,23+,24+,25-,26+,27+,28+,29+/m0/s1
SMIL COc1ccc(cc1OC)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2
Molekylvægt (g/mol) 624.59
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.