Få mere at vide
Methylamine-N,N-d2 DCl, CDN
Artikelnummer. 30321596
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30321596 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321596 Leverandør CDN Leverandørnr. CDND05795G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methylamine-N,N-D2 Deuterochloride
CAS 14779-52-3
Molekylær formel C D2 H3 N . Cl D
Mængde 5 g
Synonym Hydrochloric acid-d, compd. with methanamine-d2 (1:1) (9 CI), Hydrochloric acid-d, compd. with methylamine-d2 (1:1) (8 CI), Methanamine-d2, hydrochloride-d (9 CI), Methylamine-d2, hydrochloride-d (6 CI, 8 CI), Monomethylammonium-d3 chloride, Methylamine-d2 hydrochloride-d, Methylamine-N,N-D2 deuterochloride
InChI formel InChI=1 S/CH5N.ClH/c1-2;/h2H2,1H3;1 H/i/hD3
SMIL [2 H]Cl.[2 H]N([2 H])C
IUPAC navn (2 H)chlorane;N,N-dideuteriomethanamine
Molekylvægt (g/mol) 70.5365
Formel vægt 70.0377 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity