Få mere at vide
Methyl3-[[(2RS)-2-(Butylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Butylarticaine Hydrochloride), Mikromol™
Beskrivelse
Methyl3-[[(2RS)-2-(Butylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Butylarticaine Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl3-[((2RS)-2-(Butylamino)propanoyl)amino]-4-methylthiophene-2-carboxylate Hydrochloride (Butylarticaine Hydrochloride) |
| CAS | 23964-59-2 |
| Analyt- eller komponentnavne | Methyl3-[((2RS)-2-(Butylamino)propanoyl)amino]-4-methylthiophene-2-carboxylate Hydrochloride (Butylarticaine Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H22 N2 O3 S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Articaine Hydrochloride Imp. G (EP), Articaine Imp. G (EP), Methyl3-[[(2RS)-2-(Butylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. G (EP) as Hydrochloride, Butylarticaine Hydrochloride, Articaine Hydrochloride Impurity G as Hydrochloride, Articaine Impurity G as Hydrochloride |
| InChI formel | InChI=1S/C14H22N2O3S.ClH/c1-5-6-7-15-10(3)13(17)16-11-9(2)8-20-12(11)14(18)19-4;/h8,10,15H,5-7H2,1-4H3,(H,16,17);1H |
| SMIL | Cl.CCCCNC(C)C(=O)Nc1c(C)csc1C(=O)OC |
| IUPAC navn | methyl 3-[2-(butylamino)propanoylamino]-4-methylthiophene-2-carboxylate;hydrochloride |
| Vis mere |