Få mere at vide
Methyl Valproate, Mikromol™
Beskrivelse
Methyl Valproate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl Valproate |
| CAS | 22632-59-3 |
| Analyt- eller komponentnavne | Methyl Valproate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H18 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-Propylpentanoic acid methyl ester, Valeric acid, 2-propyl-, methyl ester (6CI,7CI,8CI), Methyl 2-propylpentanoate, Methyl dipropylacetate, Methyl valproate |
| InChI formel | InChI=1S/C9H18O2/c1-4-6-8(7-5-2)9(10)11-3/h8H,4-7H2,1-3H3 |
| SMIL | CCCC(CCC)C(=O)OC |
| IUPAC navn | methyl 2-propylpentanoate |
| Vis mere |