missing translation for 'onlineSavingsMsg'
Få mere at vide

(Methyl)triphenylphosphonium Iodide-d3, TRC

Artikelnummer. 30481069
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
Pakningsstørrelse:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30481069 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481069 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3309972.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (Methyl)triphenylphosphonium Iodide-D3
CAS 1560-56-1
Molekylær formel C19 2H3 H15 P . I
Mængde 2.5 g
Synonym Phosphonium, methyl-d3-triphenyl-, iodide (1:1) (ACI), Methyl-d3-triphenylphosphonium iodide (7CI), Phosphonium, methyl-d3-triphenyl-, iodide (8CI, 9CI), (Trideuteromethyl)triphenylphosphonium iodide, Triphenyl(trideuteriomethyl)phosphonium iodide, (Methyl)triphenylphosphonium iodide-d3, Methyltriphenylphosphonium-d3 iodide, Methyltriphenylphosphonium-D3 (methyl-D3) iodide
InChI formel InChI=1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1D3;
SMIL [I-].[2H]C([2H])([2H])[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC navn triphenyl(trideuteriomethyl)phosphanium;iodide
Molekylvægt (g/mol) 407.24
Formel vægt 407.0379
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.