Få mere at vide
Methyl Tridecanoate-d25, CDN
Artikelnummer. 30308785
Skift visning
Mængde:
0.1 g
0.5 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30308785 0.1 g 0.1g
30309968 0.5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308785 Leverandør CDN Leverandørnr. CDND58550.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Tridecanoate-D25
CAS 1219804-90-6
Molekylær formel C14 D25 H3 O2
Mængde 0.1 g
Synonym Methyl Tridecanoate-d25, Tridecanoic-d25 Acid Methyl Ester
InChI formel InChI=1 S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14(15)16-2/h3-13H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC
IUPAC navn methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-pentacosadeuteriotridecanoate
Molekylvægt (g/mol) 253.525
Formel vægt 253.3658 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity