Få mere at vide
Methyl trans-Cinnamate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl trans-Cinnamate |
| CAS | 1754-62-7 |
| Molekylær formel | C10 H10 O2 |
| Mængde | 500 g |
| Synonym | Methyl trans-Cinnamate, 2-Propenoic acid, 3-phenyl-, methyl ester, (2E)-, 2-Propenoic acid, 3-phenyl-, methyl ester, (E)-, Cinnamic acid, methyl ester, (E)- (8CI), (2E)-3-Phenyl-2-propenoic acid methyl ester, (E)-3-Phenyl-2-propenoic acid methyl ester, (E)-3-Phenylacrylic acid methyl ester, (E)-Methyl 3-phenylacrylate, (E)-Methyl cinnamate, Methyl (2E)-3-phenylpropenoate, Methyl (E)-3-phenyl-2-propenoate, Methyl (E)-3-phenylprop-2-enoate, Methyl (E)-3-phenylpropenoate, Methyl (E)-cinnamate, Methyl trans-3-phenyl-2-propenoate, trans-3-Phenyl-2-propenoic acid methyl ester, trans-Cinnamic acid methyl ester, trans-Cinnamic acid methyl ester, trans-Methyl 3-phenyl-2-propenoate, trans-Methyl cinnamate, trans-Cinnamic acid-methyl ester |
| InChI formel | InChI=1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+ |
| SMIL | COC(=O)\C=C\c1ccccc1 |
| IUPAC navn | methyl (E)-3-phenylprop-2-enoate |
| Molekylvægt (g/mol) | 162.19 |
| Formel vægt | 162.0681 |