missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl trans-6-Octadecenoate, TRC

Artikelnummer. 30135678
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135678 5 mg
30008857 50 mg
17948874 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30135678 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3245935MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 14620-36-1
Molekylær formel C19H36O2
Molekylvægt (g/mol) 296.49
Synonym Methyl trans-6-Octadecenoate
IUPAC navn methyl (E)-octadec-6-enoate
SMIL CCCCCCCCCCC\C=C\CCCCC(=O)OC

Tekniske data

Kemisk navn eller materiale Petroselaidic Acid Methyl Ester
Mængde 5 mg
InChI formel InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h13-14H,3-12,15-18H2,1-2H3/b14-13+
Formel vægt 296.2715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.