Få mere at vide
Methyl tert-butyl ether (MTBE) 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Mængde:
5 x 1 ml
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl tert-butyl ether |
| CAS | 1634-04-4 |
| Analyt- eller komponentnavne | Methyl tert-butyl ether |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C5 H12 O |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Propane, 2-methoxy-2-methyl-, Ether, tert-butyl methyl (6CI,7CI,8CI), 2-Methoxy-2-methylpropane, 1,1-Dimethylethyl methyl ether, 2-Methyl-2-methoxypropane, Driveron, MTBE, Methyl 1,1-dimethylethyl ether, Methyl tert-butyl ether, Methyl tertiary butyl ether, t-Butyl methyl ether, tert-Butoxymethane, tert-Butyl methyl ether |
| InChI formel | InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3 |
| SMIL | COC(C)(C)C |
| Vis mere |