missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Salicylate-d3, TRC

Artikelnummer. 30479138
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479138 10 mg
30468203 5 mg
30481811 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479138 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33102710MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1335435-40-9
Molekylær formel C8H5D3O3
Molekylvægt (g/mol) 155.17
Synonym 2-hydroxybenzoic Acid Methyl Ester-d3, 2-(Methoxycarbonyl)phenol-d3, 2-Carbomethoxyphenol-d3, 2-Hydroxybenzoic Acid Methyl Ester-d3, Analgit-d3, Anthrapole ND-d3, Ben Gay-d3, Exagien-d3, Flucarmit-d3, Methyl 2-Hydroxybenzoate-d3, Methyl Ester 2-Hydroxy benzoic Acid-d3, Methyl Ester of 2-hydroxybenzoic Acid-d3, Methyl o-Hydroxybenzoate-d3, NSC 8204-d3, PredaLure-d3, Wintergreen Oil-d3, o-Hydroxybenzoic Acid Methyl Ester-d3
IUPAC navn trideuteriomethyl 2-hydroxybenzoate
SMIL [2H]C([2H])([2H])OC(=O)c1ccccc1O

Tekniske data

Kemisk navn eller materiale Methyl Salicylate-d3
Mængde 10 mg
InChI formel InChI=1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3/i1D3
Formel vægt 155.0662
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.