Få mere at vide
Methyl (S)-(+)-Mandelate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl (S)-(+)-Mandelate |
| CAS | 21210-43-5 |
| Molekylær formel | C9H10O3 |
| Mængde | 5 g |
| Synonym | +)-Mandelic Acid Methyl Ester, (+)-Methyl Mandelate, (S)-(+)-Mandelic Acid Methyl Ester, (S)-(+)-Methyl Mandelate, (S)-Mandelic Acid Methyl Ester, (S)-Methyl Mandelate, L-(+)-Mandelic Acid Methyl Ester, Methyl (S)-2-Hydroxy-2-phenylacetate, Methyl (S)-alpha-Hydroxybenzeneacetate, Methyl L-Mandelate','Methyl d-Mandelate |
| InChI formel | InChI=1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3/t8-/m0/s1 |
| SMIL | COC(=O)[C@@H](O)c1ccccc1 |
| IUPAC navn | methyl (2S)-2-hydroxy-2-phenylacetate |
| Molekylvægt (g/mol) | 166.17 |
| Formel vægt | 166.063 |