missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7046387
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3976-69-0
C5H10O3
118.132
MFCD00063289
LDLDJEAVRNAEBW-SCSAIBSYSA-N
methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
2724279
methyl (3R)-3-hydroxybutanoate
CC(CC(=O)OC)O
This item is not returnable. View return policy
3976-69-0
C5H10O3
118.132
MFCD00063289
LDLDJEAVRNAEBW-SCSAIBSYSA-N
methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
2724279
methyl (3R)-3-hydroxybutanoate
CC(CC(=O)OC)O

Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral ß-lactams.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.055
Boiling Point 56°C to 58°C (11 mmHg)
Flash Point 73°C (163°F)
Odor Pleasant
Refractive Index 1.422
Quantity 1 g
Beilstein 3648161
Solubility Information Slightly soluble in water.
Optical Rotation −48.5° (c=1.3 in Chloroform)
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl (R)-(-)-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'