Få mere at vide
Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate, TRC
Artikelnummer. 30479834
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479834 250 mg
30477691 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479834 Leverandør Toronto Research Chemicals Leverandørnr. TRCM305188250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate
CAS 153294-01-0
Molekylær formel C15H23ClO3Si
Mængde 250 mg
Synonym (alphaR)-3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester, (R)- 3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester
InChI formel InChI=1S/C15H23ClO3Si/c1-15(2,3)20(5,6)19-13(14(17)18-4)11-8-7-9-12(16)10-11/h7-10,13H,1-6H3/t13-/m1/s1
SMIL COC(=O)[C@H](O[Si](C)(C)C(C)(C)C)c1cccc(Cl)c1
IUPAC navn methyl (2R)-2-[tert-butyl(dimethyl)silyl]oxy-2-(3-chlorophenyl)acetate
Molekylvægt (g/mol) 314.88
Formel vægt 314.1105