Learn More
Methyl Propionylacetate, TRC
Product Code. 30474506
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
30474506 100 g 100g
1 options
Product Code. 30474506 Supplier Toronto Research Chemicals Supplier No. TRCM326820100G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Propionylacetate
CAS 30414-53-0
Molecular Formula C6H10O3
Quantity 100 g
Synonym 3-Oxopentanoic Acid Methyl Ester, 3-oxo-Valeric Acid Methyl Ester, 3-Oxopentanoic Acid Methyl Ester, 3-Oxovaleric Acid Methyl Ester, Methyl 3-Oxopentanoate, Methyl 3-oxovalerate, Methyl Propanoylacetate, Methyl Propionylacetate, Methyl beta-Ketovalerate
InChI Formula InChI=1S/C6H10O3/c1-3-5(7)4-6(8)9-2/h3-4H2,1-2H3
SMILES CCC(=O)CC(=O)OC
IUPAC Name methyl 3-oxopentanoate
Molecular Weight (g/mol) 130.14
Formula Weight 130.063