Learn More
Methyl phenylacetate, 99+%, ACROS Orgaincs™
Brand: Acros Organics 148440010
Additional Details : CAS Number : 101-41-7 Weight : 1.11700kg
Quantity | 1 L |
---|
Chemical Identifiers
101-41-7 | |
150.18 | |
methyl 2-phenylacetate |
C9H10O2 | |
CRZQGDNQQAALAY-UHFFFAOYSA-N | |
COC(=O)CC1=CC=CC=C1 |
Specifications
Methyl phenylacetate | |
95°C | |
1.5060 to 1.5080 | |
220.0°C | |
101-41-7 | |
C9H10O2 | |
09, 434 | |
Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol | |
COC(=O)CC1=CC=CC=C1 | |
150.18 |
1.0670 g/mL | |
Glass bottle | |
1.067 | |
1 L | |
99+% | |
C6H5CH2CO2CH3 | |
15, 7380 | |
CRZQGDNQQAALAY-UHFFFAOYSA-N | |
methyl 2-phenylacetate | |
150.18 |