Få mere at vide
Methyl Paraben-d4, TRC
Artikelnummer. 30473119
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30473119 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473119 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32566325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Paraben-d4
CAS 362049-51-2
Renhedsgrad noter HPLC
Molekylær formel C8 2H4 H4 O3
Mængde 25 mg
Synonym Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, methyl ester (9CI), Methyl 4-hydroxybenzoate-2,3,5,6-D4, Methyl 4-hydroxybenzoate-d4, Methyl parahydroxybenzoate-d4, 4-Hydroxybenzoic-2,3,5,6-d4 acid methyl ester, Methylparaben-d4
InChI formel InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(C(=O)OC)c([2H])c([2H])c1O
IUPAC navn methyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 156.17
Formel vægt 156.0725
Procent renhed >95