Learn More
Thermo Scientific Chemicals Methyl orange, Indicator
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | >300.0°C |
Beilstein | 16, 331 |
Merck Index | 15, 6178 |
Solubility Information | Solubility in water: soluble in hot water. Other solubilities: practically insoluble in alcohol |
Infrared Spectrum | Authentic |
Lambda Max | 461 to 469nm (in 0.01N NaOH) |
Formula Weight | 327.32 |
Grade | Pure |
Packaging | Glass bottle |
Visual Transition Interval | from pH 3.2 (pink or red)to pH 4.4 (yellow-orange) |
Show More |
Safety and Handling
Product Identifier
- Methyl Orange
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /