Få mere at vide
Methyl Octanoate-d15, TRC
Artikelnummer. 30473504
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30473504 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473504 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3244915MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Octanoate-d15
CAS 1219798-91-0
Molekylær formel C9 D15 H3 O2
Mængde 5 mg
Synonym Methyl Octanoate-d15, Octanoic-d15 Acid Methyl Ester, Octanoic acid methyl ester-d15, Methyl caprylate-d15, Octanoic Acid Methyl Ester D15, Methyl Octanoate-D15
InChI formel InChI=1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OC
IUPAC navn methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate
Molekylvægt (g/mol) 173.3305
Formel vægt 173.2248