Få mere at vide
Methyl Octanoate-d15, CDN
Artikelnummer. 30320588
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320588 0.1 g 0.1g
30313612 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320588 Leverandør CDN Leverandørnr. CDND70050.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Octanoate-d15
CAS 1219798-91-0
Molekylær formel C9 D15 H3 O2
Mængde 0.1 g
Synonym Methyl Octanoate-d15, Octanoic-d15 Acid Methyl Ester, Octanoic acid methyl ester-d15, Methyl caprylate-d15, Octanoic Acid Methyl Ester D15, Methyl Octanoate-D15
InChI formel InChI=1 S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC
IUPAC navn methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate
Molekylvægt (g/mol) 173.3305
Formel vægt 173.2248 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity