Få mere at vide
Methyl Nicotinate-2,4,5,6-d4, TRC
Artikelnummer. 30482879
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30482879 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482879 Leverandør Toronto Research Chemicals Leverandørnr. TRCM323241100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Nicotinate-2,4,5,6-d4
CAS 345909-99-1
Molekylær formel C7 D4 H3 N O2
Mængde 100 mg
Synonym 3-Pyridine-2,4,5,6-d4-carboxylic acid','methyl ester (9CI), 3-Pyridine-2,4,5,6-d4-carboxylic acid methyl ester, Nicotinic acid-d4 methyl ester, 3-(Methoxycarbonyl)pyridine-2,4,5,6-d4, Methyl nicotinate-d4, Methyl Nicotinate-2,4,5,6-d4, Methyl Nicotinate-2,4,5,6-D4
InChI formel InChI=1S/C7H7NO2/c1-10-7(9)6-3-2-4-8-5-6/h2-5H,1H3/i2D,3D,4D,5D
SMIL [2H]c1nc([2H])c(C(=O)OC)c([2H])c1[2H]
IUPAC navn methyl 2,4,5,6-tetradeuteriopyridine-3-carboxylate
Molekylvægt (g/mol) 141.161
Formel vægt 141.073