missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Nicotinate-2,4,5,6-d4, TRC

Artikelnummer. 30481209
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481209 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481209 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32324150MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl Nicotinate-2,4,5,6-d4
CAS 345909-99-1
Molekylær formel C7 D4 H3 N O2
Mængde 50 mg
Synonym 3-Pyridine-2,4,5,6-d4-carboxylic acid','methyl ester (9CI), 3-Pyridine-2,4,5,6-d4-carboxylic acid methyl ester, Nicotinic acid-d4 methyl ester, 3-(Methoxycarbonyl)pyridine-2,4,5,6-d4, Methyl nicotinate-d4, Methyl Nicotinate-2,4,5,6-d4, Methyl Nicotinate-2,4,5,6-D4
InChI formel InChI=1S/C7H7NO2/c1-10-7(9)6-3-2-4-8-5-6/h2-5H,1H3/i2D,3D,4D,5D
SMIL [2H]c1nc([2H])c(C(=O)OC)c([2H])c1[2H]
IUPAC navn methyl 2,4,5,6-tetradeuteriopyridine-3-carboxylate
Molekylvægt (g/mol) 141.161
Formel vægt 141.073
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.