Få mere at vide
Methyl N-Acetyltyrosinate, TRC
Artikelnummer. 30009558
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009558 5 g -
30009613 1 g -
30004992 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009558 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3587455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2440-79-1
Molekylær formel C12 H15 N O4
Molekylvægt (g/mol) 237.252
Synonym 4-((2S)-2-Acetylamino-3-methoxy-3-oxopropyl)phenol, Acetyl-L-tyrosine Methyl Ester, N-Acetyltyrosine Methyl Ester
IUPAC navn methyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate
SMIL COC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)C
Tekniske data
Kemisk navn eller materiale Methyl N-Acetyltyrosinate
InChI formel InChI=1S/C12H15NO4/c1-8(14)13-11(12(16)17-2)7-9-3-5-10(15)6-4-9/h3-6,11,15H,7H2,1-2H3,(H,13,14)/t11-/m0/s1
Mængde 5 g
Formel vægt 237.1