Få mere at vide
Methyl N-Acetylglycinate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl N-Acetylglycinate |
| CAS | 1117-77-7 |
| Molekylær formel | C5 H9 N O3 |
| Mængde | 50 mg |
| Synonym | Glycine','N-acetyl-','methyl ester, Aceturic acid','methyl ester (6CI), Methyl 2-acetamidoacetate, Methyl N-acetylglycinate, Methyl N-acetylglycine, Methyl acetylglycinate, N-Acetylglycine methyl ester |
| InChI formel | InChI=1S/C5H9NO3/c1-4(7)6-3-5(8)9-2/h3H2,1-2H3,(H,6,7) |
| SMIL | COC(=O)CNC(=O)C |
| IUPAC navn | methyl 2-acetamidoacetate |
| Molekylvægt (g/mol) | 131.13 |
| Formel vægt | 131.0582 |