missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Myristate-d3, TRC

Artikelnummer. 30465299
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465299 250 mg
30472872 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465299 Leverandør Toronto Research Chemicals Leverandørnr. TRCM227602250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 60670-33-9
Molekylær formel C15H27D3O2
Molekylvægt (g/mol) 245.42
Synonym Myristic Acid Methyl Ester-d3, Emery 2214-d3, Metholeneat 2495-d3, Methyl Myristate-d3, Methyl n-Tetradecanoate-d3, Methyl Tetradecanoate-d3, NSC 5029-d3, Pastel M 14-d3, Pastell M 14-d3, Uniphat A50-d3
IUPAC navn trideuteriomethyl tetradecanoate
SMIL O=C(OC([2H])([2H])[2H])CCCCCCCCCCCCC

Tekniske data

Kemisk navn eller materiale Methyl Myristate-d3
Mængde 250 mg
InChI formel InChI=1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3/i2D3
Formel vægt 245.2434
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.