missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Mycophenolate-d6 (EP Impurity E), TRC

Artikelnummer. 30469876
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469876 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469876 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3205421MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl Mycophenolate-d6 (EP Impurity E)
CAS 1331643-25-4
Molekylær formel C18H16D6O6
Mængde 1 mg
Synonym (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic Acid Methyl Ester-d6, Mycophenolic Acid Methyl Ester-d6, EP Impurity E-d6
InChI formel InChI=1S/C18H22O6/c1-10(6-8-14(19)22-3)5-7-12-16(20)15-13(9-24-18(15)21)11(2)17(12)23-4/h5,20H,6-9H2,1-4H3/b10-5+/i3D3,4D3
SMIL [2H]C([2H])([2H])OC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC([2H])([2H])[2H])\C
IUPAC navn trideuteriomethyl (E)-6-[4-hydroxy-7-methyl-3-oxo-6-(trideuteriomethoxy)-1H-2-benzofuran-5-yl]-4-methylhex-4-enoate
Molekylvægt (g/mol) 340.4
Formel vægt 340.1793
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.