Få mere at vide
Methyl Methanethiosulfonate-13C, TRC
Artikelnummer. 30137599
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137599 100 mg
30004082 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137599 Leverandør Toronto Research Chemicals Leverandørnr. TRCM321502100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1309943-60-9
Molekylær formel C13CH6O2S2
Molekylvægt (g/mol) 127.19
Synonym Methanesulfonothioic Acid S-Methyl Ester-13C, Thiomethanesulfonic Acid S-Methyl Ester-13C, MMTS-13C, NSC 21545-13C, S-Methyl Methanesulfonothioate-13C, S-Methyl Methanethiosulfonate-13C;
SMIL O=S(S[13CH3])(C)=O
Tekniske data
Kemisk navn eller materiale Methyl-13C Methanethiosulfonate
Mængde 100 mg
InChI formel InChI=1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3/i1+1