Learn More
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals
Catalog Number p-7052640
Click to view available options
Quantity:
5 g
25 g
100 g
66-27-3
C2H6O3S
110.127
MFCD00007557
MBABOKRGFJTBAE-UHFFFAOYSA-N
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
4156
CHEBI:25255
methyl methanesulfonate
COS(=O)(=O)C
66-27-3
C2H6O3S
110.127
MFCD00007557
MBABOKRGFJTBAE-UHFFFAOYSA-N
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
4156
CHEBI:25255
methyl methanesulfonate
COS(=O)(=O)C
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.
Solubility
Very soluble in water.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3 g/mL
Boiling Point 202°C to 203°C
Flash Point 104°C (219°F)
Linear Formula CH3SO3CH3
Refractive Index 1.414
Quantity 5 g
UN Number UN2810
Beilstein 1098586
Merck Index 14,6097
Solubility Information Very soluble in water.
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl methanesulfonate