Learn More
Methyl Methacrylate, Extra Pure, SLR, Stabilised with Topanol, Fisher Chemical™
Catalog Number p-7073315
Click to view available options
Quantity:
500 mL
2.5 L
80-62-6
C5H8O2
100.117
8587
VVQNEPGJFQJSBK-UHFFFAOYSA-N
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
6658
CHEBI:34840
methyl 2-methylprop-2-enoate
CC(=C)C(=O)OC
80-62-6
C5H8O2
100.117
8587
VVQNEPGJFQJSBK-UHFFFAOYSA-N
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
6658
CHEBI:34840
methyl 2-methylprop-2-enoate
CC(=C)C(=O)OC
Melting Point -48°C
Color Colorless
Boiling Point 100°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 3.5
Formula Weight 100.12g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Methyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.