missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate, Extra Pure, SLR, Stabilised with Topanol, Fisher Chemical™

Brand:  Fisher Chemical M/4950/17

Additional Details : CAS Number : 80-62-6 Weight : 2.35000kg Transport : UN number : 1247 Chem class : 3 Pack group : II


Quantity 2.5L

 View more versions of this product

Product Code. 10459370

Quantity Price
1 364.00 DKK / 2.50 L
4 363.20 DKK / 2.50 L
16 345.04 DKK / 2.50 L
Estimated Shipment
26-05-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Chemical Identifiers

80-62-6
100.117
VVQNEPGJFQJSBK-UHFFFAOYSA-N
6658
methyl 2-methylprop-2-enoate
C5H8O2
8587
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
CHEBI:34840
CC(=C)C(=O)OC

Specifications

Amber glass bottle
100°C
-48°C
80-62-6
8587
VVQNEPGJFQJSBK-UHFFFAOYSA-N
methyl 2-methylprop-2-enoate
6658
100.12g/mol
3.5
Colorless
2.5L
C5H8O2
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
CC(=C)C(=O)OC
100.117
CHEBI:34840
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl methacrylate

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents