Få mere at vide
Methyl Margarate-d33, TRC
Artikelnummer. 30480261
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30480261 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480261 Leverandør Toronto Research Chemicals Leverandørnr. TRCM311922250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Margarate-d33
CAS 1219804-81-5
Molekylær formel C18 2H33 H3 O2
Mængde 250 mg
Synonym Heptadecanoic-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-d33 acid','methyl ester (ACI), Methyl heptadecanoate-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-d33 (ACI), Methyl Heptadecanoate-d33, Methyl Margarate-d33, Heptadecanoic Acid-d33 Methyl Ester, Heptadecanoic Acid Methyl Ester-D33 (heptadecanoic-D33)
InChI formel InChI=1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OC
IUPAC navn methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-tritriacontadeuterioheptadecanoate
Molekylvægt (g/mol) 317.68
Formel vægt 317.4787