missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl isothiocyanate, polymer-supported, 1.5-1.9 mmol/g on polystyrene, Thermo Scientific™

Artikelnummer. 11398786
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
Pakningsstørrelse:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11398786 10 g 10g
11378786 1 g 1g
11388786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11398786 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L19464.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 556-61-6
Molekylær formel C2H3NS
Molekylvægt (g/mol) 73.11
MDL nummer MFCD00004818,MFCD03457480
InChI nøgle LGDSHSYDSCRFAB-UHFFFAOYSA-N
Synonym methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide
PubChem CID 11167
ChEBI CHEBI:78337
IUPAC navn methylimino(sulfanylidene)methane
SMIL CN=C=S

Tekniske data

Kemisk navn eller materiale Methyl isothiocyanate
Navn note polymer-supported, 1.5 to 1.9mmol/g on polystyrene
Mængde 10 g
Følsomhed Moisture sensitive

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.