missing translation for 'onlineSavingsMsg'
Learn More

Methyl isonicotinate, 98%, Thermo Scientific Chemicals

Catalog Number p-164387
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
COC(=O)C1=CC=NC=C1
This item is not returnable. View return policy
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
COC(=O)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 8.5°C
Color Brown to Orange
Density 1.1610g/mL
Boiling Point 207.0°C to 209.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140
Quantity 100 mL
Beilstein 22, 46
Merck Index 10, 5034
Specific Gravity 1.161
Synonym trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
IUPAC Name trimethyl benzene-1,2,4-tricarboxylate
PubChem CID 17159
Formula Weight 137.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl isonicotinate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl isonicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'