missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Indole-3-acetate, TRC

Artikelnummer. 30467283
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467283 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30467283 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3134751G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale Methyl Indole-3-acetate
CAS 1912-33-0
Molekylær formel C11 H11 N O2
Mængde 1 g
Synonym 1H-Indole-3-acetic acid','methyl ester (9CI','ACI), Indole-3-acetic acid','methyl ester (6CI','7CI','8CI), (1H-Indol-3-yl)acetic acid methyl ester, IAA methyl ester, Indole-3-methyl acetate, Indolyl-3-acetic acid methyl ester, Methyl 1H-indol-3-ylacetate, Methyl 1H-indole-3-acetate, Methyl 2-(1H-indol-3-yl)acetate, Methyl indol-3-ylacetate, Methyl indole-3-acetate, Methyl β-indoleacetate, NSC 63806, β-Indolylacetic acid methyl ester, 1H-Indole-3-acetic Acid Methyl Ester
InChI formel InChI=1S/C11H11NO2/c1-14-11(13)6-8-7-12-10-5-3-2-4-9(8)10/h2-5,7,12H,6H2,1H3
SMIL COC(=O)Cc1c[nH]c2ccccc12
IUPAC navn methyl 2-(1H-indol-3-yl)acetate
Molekylvægt (g/mol) 189.21
Formel vægt 189.079
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.