Få mere at vide
Methyl Hydroxyethyl Piperazinyl Methyl Pyrimidinylamino Thiazolecarboxylate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl Hydroxyethyl Piperazinyl Methyl Pyrimidinylamino Thiazolecarboxylate |
| CAS | 1245157-35-0 |
| Molekylær formel | C16H22N6O3S |
| Mængde | 1 mg |
| Synonym | Methyl 2-((6-(4-(2-Hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxylate, 2-[[6-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-methyl Ester 5-Thiazolecarboxylic Acid, Methyl 2-[[6-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-5-thiazolecarboxylate |
| InChI formel | InChI=1S/C16H22N6O3S/c1-11-18-13(20-16-17-10-12(26-16)15(24)25-2)9-14(19-11)22-5-3-21(4-6-22)7-8-23/h9-10,23H,3-8H2,1-2H3,(H,17,18,19,20) |
| SMIL | COC(=O)c1cnc(Nc2cc(nc(C)n2)N3CCN(CCO)CC3)s1 |
| Molekylvægt (g/mol) | 378.45 |