missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl hydrogen adipate, 99%, Thermo Scientific™

Artikelnummer. 11362856
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11362856 25 g 25g
11352856 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11362856 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L09061.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl hydrogen adipate is a useful synthetic intermediate. It can be used to prepare 1,3,4-thiadiazole hydroxamic acid derivatives as novel histone deacetylase inhibitors which show potential antitumor activities. It can be also used to synthesize mannose-linked biphenylylacetic acid derivatives as novel inhibitors of selectin-mediated cell adhesion.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 627-91-8
Molekylær formel C7H12O4
Molekylvægt (g/mol) 160.17
MDL nummer MFCD00004418
InChI nøgle UOBSVARXACCLLH-UHFFFAOYSA-N
Synonym monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
PubChem CID 12328
ChEBI CHEBI:70855
IUPAC navn 6-methoxy-6-oxohexanoic acid
SMIL COC(=O)CCCCC(O)=O

Tekniske data

Kemisk navn eller materiale Methyl hydrogen adipate
Smeltepunkt 7°C to 9°C
Tæthed 1.122
Kogepunkt 134°C to 135°C (2 mmHg)
Flammepunkt >110°C (230°F)
Brydningsindeks 1.442
Mængde 25 g
Beilstein 1766411
Opløselighedsinformation Not miscible or difficult to mix with water.
Formel vægt 160.17
Procent renhed 99%
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.