Få mere at vide
Methyl Hexanoate-5,5,6,6,6-d5, CDN
Artikelnummer. 30325341
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325341 0.05 g -
30323165 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325341 Leverandør CDN Leverandørnr. CDND80720.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Hexanoate-5,5,6,6,6-D5
CAS 1394230-20-6
Molekylær formel C7 D5 H9 O2
Mængde 0.05 g
Synonym Hexanoic-5,5,6,6,6-d5 acid, methyl ester (ACI), Methyl hexanoate-5,5,6,6,6-d5 (ACI), Hexanoic-5,5,6,6,6-d5 acid methyl ester, Hexanoic acid methyl ester-d5, Methyl hexanoate-5,5,6,6,6-D5, Methyl hexanoate-d5
InChI formel InChI=1 S/C7H14O2/c1-3-4-5-6-7(8)9-2/h3-6H2,1-2H3/i1D3,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCC(=O)OC
IUPAC navn methyl 5,5,6,6,6-pentadeuteriohexanoate
Molekylvægt (g/mol) 135.2157
Formel vægt 135.1308 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity