Få mere at vide
Methyl Heptadecanoate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Heptadecanoic acid-methyl ester |
| CAS | 1731-92-6 |
| Molekylær formel | C18 H36 O2 |
| Mængde | 5 g |
| Synonym | Heptadecanoic acid-methyl ester, Heptadecanoic Acid Methyl Ester, Heptadecanoic acid','methyl ester (6CI','7CI','8CI','9CI','ACI), A 27667, Margaric acid methyl ester, Methyl heptadecanoate, Methyl margarate, NSC 97364 |
| InChI formel | InChI=1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3 |
| SMIL | CCCCCCCCCCCCCCCCC(=O)OC |
| IUPAC navn | methyl heptadecanoate |
| Molekylvægt (g/mol) | 284.48 |
| Formel vægt | 284.2715 |