Learn More
Methyl formate, 98%, for spectroscopy, Thermo Scientific Chemicals
Catalog Number p-163371
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
7865
CHEBI:77699
methyl formate
COC=O
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
7865
CHEBI:77699
methyl formate
COC=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Acidity 0.0008 meq/g max.
Absorbance 0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
Melting Point -100.0°C
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HCO2CH3
Refractive Index 1.3420 to 1.3440
Quantity 5 mL
Beilstein 02, 18
Residue after Evaporation 0.0005% max.
Fieser 05,446
Merck Index 15, 6148
Specific Gravity 0.968
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 98%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Methyl formate, 98%