Få mere at vide
Methyl Formate-d4, CDN
Artikelnummer. 30312380
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312380 1 g
30309471 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312380 Leverandør CDN Leverandørnr. CDND00241G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Formate-D4
CAS 23731-40-0
Molekylær formel C2 D4 O2
Mængde 1 g
Synonym Formic-d acid, methyl-d3 ester (8 CI, 9 CI), Methan-d3-ol, formate-d (8 CI), Methyl Formate-D4, Methyl formate-d4, Methyl-d3 formate-d, Formic-d acid methyl-d3 ester, Formic acid methyl ester-d4
InChI formel InChI=1 S/C2H4O2/c1-4-2-3/h2H,1H3/i1D3,2 D
SMIL [2 H]C(=O)OC([2 H])([2 H])[2 H]
IUPAC navn trideuteriomethyl deuterioformate
Molekylvægt (g/mol) 64.0766
Formel vægt 64.0462 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity