Få mere at vide
Methyl Flurbiprofen, TRC
Artikelnummer. 30470013
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470013 250 mg
30467508 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30470013 Leverandør Toronto Research Chemicals Leverandørnr. TRCM305350250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66202-86-6
Molekylær formel C16 H15 F O2
Molekylvægt (g/mol) 258.29
Synonym 2-Fluoro-α-methyl-[1,1'-biphenyl]-4-acetic acid methyl ester, (±)-Flubiprofen methyl ester, Flurbiprofen methyl ester, Methyl flurbiprofen
IUPAC navn methyl 2-(3-fluoro-4-phenylphenyl)propanoate
SMIL COC(=O)C(C)c1ccc(c(F)c1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Methyl Flurbiprofen
Mængde 250 mg
InChI formel InChI=1S/C16H15FO2/c1-11(16(18)19-2)13-8-9-14(15(17)10-13)12-6-4-3-5-7-12/h3-11H,1-2H3
Formel vægt 258.1056