Learn More
α-Methyl-DL-phenylalanine, 98%, Thermo Scientific™
Catalog Number p-4323412
Click to view available options
Quantity:
1 g
5 g
1132-26-9
C10H13NO2
179.22
MFCD00010512
IRZQDMYEJPNDEN-UHFFFAOYNA-N
alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
108055
2-amino-2-methyl-3-phenylpropanoic acid
CC(C(N)C(O)=O)C1=CC=CC=C1
1132-26-9
C10H13NO2
179.22
MFCD00010512
IRZQDMYEJPNDEN-UHFFFAOYNA-N
alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
108055
2-amino-2-methyl-3-phenylpropanoic acid
CC(C(N)C(O)=O)C1=CC=CC=C1
Melting Point 284°C to 286°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 179.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α-Methyl-DL-phenylalanine