Få mere at vide
Methyl Dimethylbenzeneacetate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl Dimethylbenzeneacetate |
| CAS | 57625-74-8 |
| Molekylær formel | C11H14O2 |
| Mængde | 2.5 g |
| Synonym | 2-Methyl-2-phenylpropanoic Acid Methyl Ester, 2-Methyl-2-phenylpropionic Acid Methyl Ester, Methyl 2,2-dimethyl-2-phenylacetate, Methyl 2-Methyl-2-phenylpropanoate, Methyl 2-Methyl-2-phenylpropionate, Methyl alpha,alpha-Dimethylphenylacetate |
| InChI formel | InChI=1S/C11H14O2/c1-11(2,10(12)13-3)9-7-5-4-6-8-9/h4-8H,1-3H3 |
| SMIL | COC(=O)C(C)(C)c1ccccc1 |
| IUPAC navn | methyl 2-methyl-2-phenylpropanoate |
| Molekylvægt (g/mol) | 178.23 |
| Formel vægt | 178.0994 |