missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl Diethyldithiocarbamate-d3, TRC

Artikelnummer. 30465948
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465948 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465948 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30170250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl Diethyldithiocarbamate-d3
CAS 1246816-30-7
Molekylær formel C6H10D3NS2
Mængde 50 mg
Synonym N,N-Diethylcarbamodithioic Acid Methyl-d3 Ester, Diethyldithiocarbamic Acid Methyl-d3 Ester, Diethyldithiocarbamate Methyl-d3 Ester, NSC 133269-d3
InChI formel InChI=1S/C6H13NS2/c1-4-7(5-2)6(8)9-3/h4-5H2,1-3H3/i3D3
SMIL S=C(SC([2H])([2H])[2H])N(CC)CC
Molekylvægt (g/mol) 166.32
Formel vægt 166.0678
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.