Learn More
Methyl dichloromethoxyacetate, 90%, Tech., Thermo Scientific™
Catalog Number 10023130
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
CAS 17640-25-4
Color Colorless
Density 1.3400g/mL
Boiling Point 181.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C4H6Cl2O3
Refractive Index 1.4496 to 1.4516
MDL Number MFCD00012202
Quantity 25 mL
Specific Gravity 1.34
Synonym methyldichloromethoxyacetate, methyl2,2-dichloro-2-methoxyacetate, methoxydichloroacetic acid methyl ester, methyl 2,2-dichloro-2-methoxyacetate, methyl 2 pound not2-dichloro-2-methoxyacetate, aceticacid,2,2-dichloro-2-methoxy-,methylester, acetic acid,2,2-dichloro-2-methoxy-,methyl ester
InChI Key FWIGIASHGJFYOL-UHFFFAOYSA-N
SMILES COC(=O)C(OC)(Cl)Cl
IUPAC Name methyl 2,2-dichloro-2-methoxyacetate
Molecular Weight (g/mol) 173
PubChem CID 2724502
Formula Weight 173
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl dichloromethoxyacetate, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl dichloromethoxyacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.