missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl-d3-triphenylphosphonium Bromide, TRC

Artikelnummer. 30473248
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473248 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473248 Leverandør Toronto Research Chemicals Leverandørnr. TRCM331181100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl-d3-triphenylphosphonium Bromide
CAS 1787-44-6
Molekylær formel C19 D3 H15 P . Br
Mængde 100 mg
Synonym Phosphonium, methyl-d3-triphenyl-, bromide (1:1) (ACI), Methyl-d3-triphenylphosphonium bromide (7CI), Phosphonium, methyl-d3-triphenyl-, bromide (8CI, 9CI), Trideuteriomethyltriphenylphosphonium bromide, (Trideuteromethyl)triphenylphosphonium bromide, Triphenyl(trideuteriomethyl)phosphonium bromide, (Methyl)triphenylphosphonium bromide-d3, Methyltriphenylphosphonium-d3 bromide, Methyltriphenylphosphonium-D3 (methyl-D3) bromide
InChI formel InChI=1S/C19H18P.BrH/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1D3;
SMIL [Br-].[2H]C([2H])([2H])[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC navn triphenyl(trideuteriomethyl)phosphanium;bromide
Molekylvægt (g/mol) 360.243
Formel vægt 359.052
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.