Få mere at vide
Methyl-d3 Toluenesulfonate, TRC
Artikelnummer. 30471245
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471245 250 mg
30467653 1 g
30468839 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471245 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330667250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7575-93-1
Molekylær formel C8 D3 H7 O3 S
Molekylvægt (g/mol) 189.247
Synonym Methan-d3-ol, 4-methylbenzenesulfonate (9CI, ACI), Methan-d3-ol, p-toluenesulfonate (8CI), p-Toluenesulfonic acid, methyl-d3 ester (6CI, 7CI, 8CI), Methyl-d3 4-methylbenzenesulfonate, Methyl-d3 tosylate, Trideuteriomethyl 4-methoxybenzenesulfonate, Trideuteriomethyl 4-methylbenzenesulfonate, Trideuteriomethyl p-toluenesulfonate, Trideuteromethyl tosylate, Methyl-d3 p-Toluenesulfonate, Methyl Toluene-4-sulfonate-D3 (methoxy-D3)
IUPAC navn trideuteriomethyl 4-methylbenzenesulfonate
SMIL [2H]C([2H])([2H])OS(=O)(=O)c1ccc(C)cc1
Tekniske data
Kemisk navn eller materiale Methyl-d3 Toluenesulfonate
Mængde 250 mg
InChI formel InChI=1S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6H,1-2H3/i2D3
Formel vægt 189.054